Gene Transcripts

Gene Symbol
Crbn
Entrez Gene ID
297498
Gene Name
cereblon
Gene Aliases
Chromosome Location
Chr. 4: 138866761 - 138885556 on Build Rnor_6.0
UniGene ID
Rn.195259
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015003.2 NP_001015003.1 5 564
GenBank mRNA AY973953.1 - 5 551
BC099779.1 - 5 549
BC127455.1 - 5 556 7112865

View Full Product Details