Gene Transcripts

Gene Symbol
Als2cr12
Entrez Gene ID
316413
Gene Name
amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 12
Gene Aliases
Chromosome Location
Chr. 9: 65665663 - 65693822 on Build Rnor_6.0
UniGene ID
Rn.195901
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014101.1 NP_001014123.1 2, 3 482
GenBank mRNA BC079241.1 - 2, 3 482

View Full Product Details