Gene Transcripts

Gene Symbol
LRRC66
Entrez Gene ID
339977
Gene Name
leucine rich repeat containing 66
Gene Aliases
Chromosome Location
Chr. 4: 51993640 - 52020311 on Build GRCh38
UniGene ID
Hs.661450
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024611.2 NP_001019782.1 4 3147
XM_011534383.2 XP_011532685.1 5 4868
XM_017008162.1 XP_016863651.1 3 6259
GenBank mRNA CR749811.1 - 4 3147

View Full Product Details