Gene Transcripts

Gene Symbol
ARHGEF35
Entrez Gene ID
445328
Gene Name
Rho guanine nucleotide exchange factor 35
Gene Aliases
ARHGEF5L
Chromosome Location
Chr. 7: 144186083 - 144195698 on Build GRCh38
UniGene ID
Hs.534621
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003702.2 NP_001003702.2 2 1812
GenBank mRNA AK125680.1 - 2 1757
BC136868.1 - 2 1671 9020859
BC136878.1 - 2 1671 9020869
DQ786206.1 - 1 430
EF560743.1 - 2 1691

View Full Product Details