Gene Transcripts

Gene Symbol
Cyp2j11
Entrez Gene ID
100066
Gene Name
cytochrome P450, family 2, subfamily j, polypeptide 11
Gene Aliases
AI314783, Cyp2j11-ps
Chromosome Location
Chr. 4: 96294508 - 96348697 on Build GRCm38
UniGene ID
Mm.26915
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004141.2 NP_001004141.2 8 1335
GenBank mRNA AK143983.1 - 8 1317
BC080831.1 - 8 1311 30469795

View Full Product Details