Gene Transcripts

Gene Symbol
OR10K2
Entrez Gene ID
391107
Gene Name
olfactory receptor family 10 subfamily K member 2
Gene Aliases
OR1-4
Chromosome Location
Chr. 1: 158419928 - 158420866 on Build GRCh38
UniGene ID
Hs.626815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004476.1 NP_001004476.1 1 573

View Full Product Details