Gene Transcripts

Gene Symbol
OR4K15
Entrez Gene ID
81127
Gene Name
olfactory receptor family 4 subfamily K member 15
Gene Aliases
OR14-20, OR4K15Q
Chromosome Location
Chr. 14: 19975519 - 19976565 on Build GRCh38
UniGene ID
Hs.554583
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005486.1 NP_001005486.1 1 978
GenBank mRNA BC140726.1 - 1 1053

View Full Product Details