Gene Transcripts

Gene Symbol
TXLNGY
Entrez Gene ID
246126
Gene Name
taxilin gamma pseudogene, Y-linked
Gene Aliases
CYorf15A, CYorf15B, TXLNG2P
Chromosome Location
Chr. Y: 19567358 - 19607170 on Build GRCh38
UniGene ID
Hs.522863
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK090535.1 - 1 1755
AK127387.1 - 3 1468
BC110817.1 - 4 1487 6058816

View Full Product Details