Gene Transcripts

Gene Symbol
BTBD2
Entrez Gene ID
55643
Gene Name
BTB domain containing 2
Gene Aliases
Chromosome Location
Chr. 19: 1985438 - 2015703 on Build GRCh38
UniGene ID
Hs.465543
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017797.3 NP_060267.2 6, 7 1189
XM_005259593.1 XP_005259650.1 6, 7 954
XM_011528127.1 XP_011526429.1 4, 5 555
GenBank mRNA AF355797.1 - 6, 7 1189
AK000393.1 - 3, 4 983
AK027481.1 - 4, 5 551
AK056818.1 - 1 1324
AK225831.1 - 6, 7 1188
AL360275.1 - 3, 4 420
AY007116.1 - 6, 7 911
BC000564.2 - 3, 4 479
BC008035.2 - 3, 4 473
BC053986.1 - 2, 3 374
BC069199.1 - 4, 5 615
BC094820.1 - 6, 7 923
CR457230.1 - 2, 3 226

View Full Product Details