Gene Transcripts

Gene Symbol
Abhd14b
Entrez Gene ID
300983
Gene Name
abhydrolase domain containing 14b
Gene Aliases
Chromosome Location
Chr. 8: 115149726 - 115154556 on Build Rnor_6.0
UniGene ID
Rn.2080
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007664.1 NP_001007665.1 4 1081
XM_006243724.1 XP_006243786.1 3 1022
GenBank mRNA BC076385.1 - 4 1081

View Full Product Details