Gene Transcripts

Gene Symbol
Vom1r5
Entrez Gene ID
494246
Gene Name
vomeronasal 1 receptor 5
Gene Aliases
V1re27
Chromosome Location
Chr. 1: 86672176 - 86673081 on Build Rnor_6.0
UniGene ID
Rn.129460
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009526.1 NP_001009526.1 1 304
GenBank mRNA AY510322.1 - 1 304

View Full Product Details