Gene Transcripts

Gene Symbol
C1QL3
Entrez Gene ID
389941
Gene Name
complement C1q like 3
Gene Aliases
C1QTNF13, C1ql, CTRP13, K100
Chromosome Location
Chr. 10: 16513743 - 16522005 on Build GRCh38
UniGene ID
Hs.676792
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010908.1 NP_001010908.1 2 1755
GenBank mRNA AK094686.1 - 1 36
BC127716.1 - 3 823 40127920
BC127717.1 - 3 769 40127921

View Full Product Details