Gene Transcripts

Gene Symbol
Fam89a
Entrez Gene ID
361441
Gene Name
family with sequence similarity 89, member A
Gene Aliases
RGD1309879
Chromosome Location
Chr. 19: 57502308 - 57514896 on Build Rnor_6.0
UniGene ID
Rn.66325
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011711.1 NP_001011711.1 2 345
GenBank mRNA AY569012.1 - 2 345
BC107921.1 - 2 385 7445343

View Full Product Details