Gene Transcripts

Gene Symbol
Sf3b4
Entrez Gene ID
295270
Gene Name
splicing factor 3b, subunit 4
Gene Aliases
Chromosome Location
Chr. 2: 198312428 - 198317180 on Build Rnor_6.0
UniGene ID
Rn.116275
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011951.1 NP_001011951.1 6 1520
GenBank mRNA BC078997.1 - 6 1520

View Full Product Details