Gene Transcripts

Gene Symbol
Rsph1
Entrez Gene ID
361818
Gene Name
radial spoke head 1 homolog (Chlamydomonas)
Gene Aliases
Tsga2
Chromosome Location
Chr. 20: 9998698 - 10020719 on Build Rnor_6.0
UniGene ID
Rn.14627
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012176.1 NP_001012176.1 3 377
GenBank mRNA BC084704.1 - 3 377

View Full Product Details