Gene Transcripts

Gene Symbol
Trim44
Entrez Gene ID
362172
Gene Name
tripartite motif-containing 44
Gene Aliases
Chromosome Location
Chr. 3: 91968781 - 92242138 on Build Rnor_6.0
UniGene ID
Rn.229767
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013203.1 NP_001013221.1 3 902
GenBank mRNA AY551091.1 - 3 902

View Full Product Details