Gene Transcripts

Gene Symbol
Ttc4
Entrez Gene ID
362556
Gene Name
tetratricopeptide repeat domain 4
Gene Aliases
Chromosome Location
Chr. 5: 126264758 - 126282429 on Build Rnor_6.0
UniGene ID
Rn.213644
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013214.1 NP_001013232.1 6 1599
GenBank mRNA BC083794.1 - 6 1599

View Full Product Details