Gene Transcripts
- Gene Symbol
- HMCN2
- Entrez Gene ID
- 256158
- Gene Name
- hemicentin 2
- Gene Aliases
- Chromosome Location
- Chr. 9: 130265742 - 130434123 on Build GRCh38
- UniGene ID
- Hs.32194
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128020.1 |
- |
11 |
2115 |
|
View Full Product Details