Gene Transcripts

Gene Symbol
Fbxw24
Entrez Gene ID
382106
Gene Name
F-box and WD-40 domain protein 24
Gene Aliases
EG382106, Gm5162
Chromosome Location
Chr. 9: 109601116 - 109626059 on Build GRCm38
UniGene ID
Mm.441401
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013776.4 NP_001013798.2 9 1274
GenBank mRNA BC080701.1 - 9 1275
CO808277.1 - 4 622
DV651634.1 - 5 712
GQ418172.1 - 9 1275

View Full Product Details