Gene Transcripts

Gene Symbol
l7Rn6
Entrez Gene ID
293103
Gene Name
lethal, Chr 7, Rinchik 6
Gene Aliases
Chromosome Location
Chr. 1: 154147098 - 154170428 on Build Rnor_6.0
UniGene ID
Rn.96145
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013897.1 NP_001013919.1 6 913
XM_006229662.2 XP_006229724.1 5 880
XM_008759638.1 XP_008757860.1 5 711
XM_008759639.1 XP_008757861.1 4 640
GenBank mRNA BC088340.1 - 6 913
FQ212130.1 - 5 824
FQ213824.1 - 5 825
FQ214677.1 - 5 825
FQ215506.1 - 5 816
FQ216197.1 - 5 825
FQ216668.1 - 5 825
FQ216759.1 - 5 825
FQ218863.1 - 5 822
FQ220044.1 - 5 825
FQ220310.1 - 5 876
FQ220575.1 - 5 825
FQ224272.1 - 5 827
FQ224945.1 - 5 825
FQ229506.1 - 5 821

View Full Product Details