Gene Transcripts

Gene Symbol
Hdhd2
Entrez Gene ID
361351
Gene Name
haloacid dehalogenase-like hydrolase domain containing 2
Gene Aliases
Ier3ip1, LRRG00122, RGD1308579
Chromosome Location
Chr. 18: 73226630 - 73278062 on Build Rnor_6.0
UniGene ID
Rn.7319
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006254965.2 XP_006255027.1 7 1007
XM_006254966.2 XP_006255028.1 7 1460
GenBank mRNA BC078941.1 - 7 981

View Full Product Details