Gene Transcripts

Gene Symbol
Dnaaf1
Entrez Gene ID
361419
Gene Name
dynein, axonemal, assembly factor 1
Gene Aliases
Lrrc50, RGD1310542
Chromosome Location
Chr. 19: 52225582 - 52245930 on Build Rnor_6.0
UniGene ID
Rn.160929
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014154.1 NP_001014176.1 11 1662
GenBank mRNA BC079038.1 - 11 1662

View Full Product Details