Gene Transcripts

Gene Symbol
Cnrip1
Entrez Gene ID
364208
Gene Name
cannabinoid receptor interacting protein 1
Gene Aliases
Crip1a, RGD1308373
Chromosome Location
Chr. 14: 100217547 - 100247235 on Build Rnor_6.0
UniGene ID
Rn.8173
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014232.1 NP_001014254.1 3 709
GenBank mRNA AY883936.1 - 3 798
BC088754.1 - 3 709 7319918

View Full Product Details