Gene Transcripts

Gene Symbol
Crls1
Entrez Gene ID
366196
Gene Name
cardiolipin synthase 1
Gene Aliases
RGD1311037
Chromosome Location
Chr. 3: 125503638 - 125522446 on Build Rnor_6.0
UniGene ID
Rn.35982
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014258.1 NP_001014280.1 1, 2 438
GenBank mRNA BC085849.1 - 1, 2 438

View Full Product Details