Gene Transcripts

Gene Symbol
Lrrc1
Entrez Gene ID
367113
Gene Name
leucine rich repeat containing 1
Gene Aliases
Chromosome Location
Chr. 8: 84721621 - 84835074 on Build Rnor_6.0
UniGene ID
Rn.15622
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014268.1 NP_001014290.1 6 628
GenBank mRNA BC079423.1 - 6 628

View Full Product Details