Gene Transcripts

Gene Symbol
Slco6d1
Entrez Gene ID
367321
Gene Name
solute carrier organic anion transporter family, member 6d1
Gene Aliases
Chromosome Location
Chr. 9: 104756808 - 104870397 on Build Rnor_6.0
UniGene ID
Rn.112361
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014270.1 NP_001014292.1 363
XM_006245580.2 XP_006245642.1 526
GenBank mRNA BC083829.1 - 363

View Full Product Details