Gene Transcripts

Gene Symbol
Armcx2
Entrez Gene ID
367903
Gene Name
armadillo repeat containing, X-linked 2
Gene Aliases
Chromosome Location
Chr. X: 105617383 - 105622212 on Build Rnor_6.0
UniGene ID
Rn.15754
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014274.1 NP_001014296.1 5 2473
XM_008773429.1 XP_008771651.1 5 2583
GenBank mRNA BC081886.1 - 5 2473

View Full Product Details