Gene Transcripts

Gene Symbol
Rab24
Entrez Gene ID
361208
Gene Name
RAB24, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 17: 9837259 - 9839464 on Build Rnor_6.0
UniGene ID
Rn.6295
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015023.1 NP_001015023.1 6 1136
XM_006253632.2 XP_006253694.1 7 835
GenBank mRNA BC091184.1 - 6 1136

View Full Product Details