Gene Transcripts

Gene Symbol
VMAC
Entrez Gene ID
400673
Gene Name
vimentin-type intermediate filament associated coiled-coil protein
Gene Aliases
Chromosome Location
Chr. 19: 5904841 - 5910252 on Build GRCh38
UniGene ID
Hs.620821
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017921.3 NP_001017921.1 1 82
GenBank mRNA AK124162.1 - 1 49
BC110802.1 - 1 49 5108486
DA541168.1 - 1 82

View Full Product Details