Gene Transcripts

Gene Symbol
Dram2
Entrez Gene ID
362011
Gene Name
DNA-damage regulated autophagy modulator 2
Gene Aliases
Tmem77
Chromosome Location
Chr. 2: 209160904 - 209189641 on Build Rnor_6.0
UniGene ID
Rn.1279
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025018.1 NP_001020189.1 10 1197
XM_006233120.2 XP_006233182.1 10 1141
XM_006233121.2 XP_006233183.1 9 1081
XM_006233122.2 XP_006233184.1 9 1182
XM_006233123.2 XP_006233185.1 9 1044
GenBank mRNA BC091251.1 - 10 1197
FQ228782.1 - 10 1294

View Full Product Details