Gene Transcripts

Gene Symbol
Fundc1
Entrez Gene ID
363442
Gene Name
FUN14 domain containing 1
Gene Aliases
Chromosome Location
Chr. X: 5549468 - 5566125 on Build Rnor_6.0
UniGene ID
Rn.22678
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025027.1 NP_001020198.1 2 195
GenBank mRNA BC091353.1 - 2 195

View Full Product Details