Gene Transcripts

Gene Symbol
Tomm6
Entrez Gene ID
66119
Gene Name
translocase of outer mitochondrial membrane 6 homolog (yeast)
Gene Aliases
1110002E23Rik, AI663970
Chromosome Location
Chr. 17: 47686645 - 47688386 on Build GRCm38
UniGene ID
Mm.89912
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164729.1 NP_001158201.1 3 633
GenBank mRNA BC021590.1 - 2 763
BC089325.1 - 1 1427
CA319049.1 - 3 282

View Full Product Details