Gene Transcripts

Gene Symbol
Ociad2
Entrez Gene ID
433904
Gene Name
OCIA domain containing 2
Gene Aliases
1810027I20Rik
Chromosome Location
Chr. 5: 73322199 - 73339355 on Build GRCm38
UniGene ID
Mm.274892
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026950.3 NP_081226.1 7 1027
GenBank mRNA BC044883.1 - 7 940

View Full Product Details