Gene Transcripts

Gene Symbol
Klri1
Entrez Gene ID
503651
Gene Name
killer cell lectin-like receptor family I member 1
Gene Aliases
Chromosome Location
Chr. 4: 163494979 - 163528237 on Build Rnor_6.0
UniGene ID
Rn.134172
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012649.1 NP_001012667.1 2 229
XM_008763328.1 XP_008761550.1 2 298
GenBank mRNA AY286494.1 - 2 229
FQ233804.1 - 2 247

View Full Product Details