Gene Transcripts

Gene Symbol
Gstm4
Entrez Gene ID
499689
Gene Name
glutathione S-transferase mu 4
Gene Aliases
Chromosome Location
Chr. 2: 210834018 - 210839242 on Build Rnor_6.0
UniGene ID
Rn.198607
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024304.1 NP_001019475.1 8 1211
GenBank mRNA BC091199.1 - 8 1211
FQ223925.1 - 7 1073

View Full Product Details