Gene Transcripts
- Gene Symbol
- VPS13B
- Entrez Gene ID
- 157680
- Gene Name
- vacuolar protein sorting 13 homolog B
- Gene Aliases
- CHS1, COH1
- Chromosome Location
- Chr. 8: 99007014 - 99877586 on Build GRCh38
- UniGene ID
- Hs.191540
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015243.2 |
NP_056058.2
|
2, 3 |
266 |
|
NM_017890.4 |
NP_060360.3
|
2, 3 |
266 |
|
NM_152564.4 |
NP_689777.3
|
2, 3 |
266 |
|
NM_181661.2 |
NP_858047.2
|
2, 3 |
266 |
|
NR_047582.1 |
|
2, 3 |
266 |
|
XM_005250800.3 |
XP_005250857.1
|
2, 3 |
572 |
|
XM_005250801.4 |
XP_005250858.1
|
2, 3 |
6442 |
|
XM_006716510.3 |
XP_006716573.1
|
2, 3 |
258 |
|
XM_011516848.2 |
XP_011515150.1
|
2, 3 |
258 |
|
XM_011516849.2 |
XP_011515151.1
|
2, 3 |
258 |
|
XM_011516853.2 |
XP_011515155.1
|
2, 3 |
258 |
|
XM_011516859.2 |
XP_011515161.1
|
2, 3 |
258 |
|
XM_011516861.1 |
XP_011515163.1
|
2, 3 |
258 |
|
XM_011516865.1 |
XP_011515167.1
|
2, 3 |
258 |
|
XM_011516866.2 |
XP_011515168.1
|
2, 3 |
258 |
|
XM_017013109.1 |
XP_016868598.1
|
2, 3 |
258 |
|
GenBank mRNA |
AJ608772.1 |
- |
2, 3 |
258 |
|
AJ608773.1 |
- |
2, 3 |
258 |
|
AK000590.1 |
- |
2, 3 |
292 |
|
AK223122.1 |
- |
2, 3 |
292 |
|
AY223814.1 |
- |
2, 3 |
266 |
|
AY223815.1 |
- |
2, 3 |
266 |
|
AY223816.1 |
- |
2, 3 |
266 |
|
AY223817.1 |
- |
2, 3 |
266 |
|
AY223818.1 |
- |
2, 3 |
266 |
|
BC094749.1 |
- |
2, 3 |
222 |
|
BC132976.1 |
- |
1, 2 |
189 |
|
BC132978.1 |
- |
1, 2 |
189 |
|
View Full Product Details