Gene Transcripts

Gene Symbol
Efcab11
Entrez Gene ID
500705
Gene Name
EF-hand calcium binding domain 11
Gene Aliases
Chromosome Location
Chr. 6: 123742971 - 123894816 on Build Rnor_6.0
UniGene ID
Rn.119809
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024350.1 NP_001019521.1 2 189
XM_006240446.2 XP_006240508.2 2 291
GenBank mRNA BC079408.1 - 2 189

View Full Product Details