Gene Transcripts

Gene Symbol
H1fnt
Entrez Gene ID
500928
Gene Name
H1 histone family, member N, testis-specific
Gene Aliases
Chromosome Location
Chr. 7: 139871491 - 139872858 on Build Rnor_6.0
UniGene ID
Rn.123328
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024356.1 NP_001019527.1 1 763
GenBank mRNA BC085950.1 - 1 763

View Full Product Details