Gene Transcripts
- Gene Symbol
- Tc2n
- Entrez Gene ID
- 500707
- Gene Name
- tandem C2 domains, nuclear
- Gene Aliases
- Mtac2d1, Tac2-N
- Chromosome Location
- Chr. 6: 125437767 - 125601408 on Build Rnor_6.0
- UniGene ID
- Rn.43919
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001025152.1 |
NP_001020323.1
|
7, 8 |
846 |
|
XM_008764831.1 |
XP_008763053.1
|
6, 7 |
2509 |
|
XM_008764832.1 |
XP_008763054.1
|
7, 8 |
2158 |
|
XM_008764833.1 |
XP_008763055.1
|
7, 8 |
2405 |
|
XM_008764834.1 |
XP_008763056.1
|
6, 7 |
1379 |
|
XM_008764835.1 |
XP_008763057.1
|
7, 8 |
1410 |
|
XM_008764836.1 |
XP_008763058.1
|
7, 8 |
1028 |
|
XM_008764837.1 |
XP_008763059.1
|
7, 8 |
1373 |
|
XM_008764838.1 |
XP_008763060.1
|
6, 7 |
1342 |
|
XM_008764839.1 |
XP_008763061.1
|
6, 7 |
908 |
|
XM_008764840.1 |
XP_008763062.1
|
5, 6 |
775 |
|
GenBank mRNA |
BC088432.1 |
- |
7, 8 |
846 |
|
FQ211170.1 |
- |
7, 8 |
964 |
|
View Full Product Details