Gene Transcripts

Gene Symbol
Hist1h3a
Entrez Gene ID
360198
Gene Name
histone cluster 1, H3a
Gene Aliases
Chromosome Location
Chr. 13: 23761885 - 23762386 on Build GRCm38
UniGene ID
Mm.221301
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK018952.1 - 1 459

View Full Product Details