Gene Transcripts

Gene Symbol
Cyp2c67
Entrez Gene ID
545288
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 67
Gene Aliases
C730004C24Rik
Chromosome Location
Chr. 19: 39608844 - 39649083 on Build GRCm38
UniGene ID
Mm.297048
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024719.2 NP_001019890.1 7 1082
GenBank mRNA AK149389.1 - 7 1093
BC092260.1 - 7 1082 5132591
BC106142.1 - 7 1082 5051946

View Full Product Details