Gene Transcripts

Gene Symbol
YOD1
Entrez Gene ID
55432
Gene Name
YOD1 deubiquitinase
Gene Aliases
DUBA8, OTUD2, PRO0907
Chromosome Location
Chr. 1: 207043849 - 207052980 on Build GRCh38
UniGene ID
Hs.567533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276320.1 NP_001263249.1 4 5954
NM_018566.3 NP_061036.3 2 5928
GenBank mRNA BC137166.1 - 2 5921
BC137167.1 - 2 5922 9021164

View Full Product Details