Gene Transcripts

Gene Symbol
Gphb5
Entrez Gene ID
366668
Gene Name
glycoprotein hormone beta 5
Gene Aliases
Gpb5
Chromosome Location
Chr. 6: 98380500 - 98383049 on Build Rnor_6.0
UniGene ID
Rn.127689
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007013.1 NP_001007014.1 1 14
GenBank mRNA AY437504.1 - 1 14

View Full Product Details