Gene Transcripts

Gene Symbol
Prss46
Entrez Gene ID
408245
Gene Name
protease, serine, 46
Gene Aliases
Tessp6
Chromosome Location
Chr. 8: 119121671 - 119133455 on Build Rnor_6.0
UniGene ID
Rn.129743
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008865.1 NP_001008865.1 2 284
XM_006243993.2 XP_006244055.1 3 524

View Full Product Details