Gene Transcripts

Gene Symbol
Yod1
Entrez Gene ID
363982
Gene Name
YOD1 deubiquitinase
Gene Aliases
hshin7
Chromosome Location
Chr. 13: 47441296 - 47443013 on Build Rnor_6.0
UniGene ID
Rn.212904
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008889.1 NP_001008889.1 2 633
GenBank mRNA BC107904.1 - 2 1126

View Full Product Details