Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026890.2 | NP_081166.1 | 3 | 231 | |
GenBank mRNA | AK005098.1 | - | 3 | 113 | |
BC023770.1 | - | 2 | 116 | ||
BC052790.1 | - | 3 | 169 | 1515292 | |
CN715203.1 | - | 3 | 231 | ||
EF437387.1 | - | 3 | 120 |