Gene Transcripts

Gene Symbol
Rab28
Entrez Gene ID
100972
Gene Name
RAB28, member RAS oncogene family
Gene Aliases
2700023P08Rik, AW496496
Chromosome Location
Chr. 5: 41624974 - 41708192 on Build GRCm38
UniGene ID
Mm.41555
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027295.2 NP_081571.1 7 1598
GenBank mRNA AK012286.1 - 7 827
AK159493.1 - 7 1599
BC004580.1 - 7 1598 3587008

View Full Product Details