Gene Transcripts

Gene Symbol
Pex1
Entrez Gene ID
71382
Gene Name
peroxisomal biogenesis factor 1
Gene Aliases
5430414H02Rik, E330005K07Rik, ZWS1
Chromosome Location
Chr. 5: 3596066 - 3637230 on Build GRCm38
UniGene ID
Mm.379196
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293806.1 NP_001280735.1 11, 12 2224
NM_027777.2 NP_082053.1 10, 11 2104
GenBank mRNA BC090845.1 - 10, 11 2104

View Full Product Details