Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018000.2 | NP_060470.2 | 3 | 601 | |
XM_011511468.2 | XP_011509770.1 | 4 | 601 | ||
GenBank mRNA | AF068290.1 | - | 2 | 100 | |
AK000978.1 | - | 3 | 571 | ||
BC032747.1 | - | 3 | 610 | 5573149 | |
BC082990.1 | - | 3 | 591 | 6159624 | |
CX781801.1 | - | 3 | 596 |