Gene Transcripts

Gene Symbol
Slpil3
Entrez Gene ID
408224
Gene Name
antileukoproteinase-like 3
Gene Aliases
Chromosome Location
Chr. 3: 160774867 - 160777100 on Build Rnor_6.0
UniGene ID
Rn.198175
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008873.1 NP_001008873.1 2 218
XM_006235602.2 XP_006235664.1 2 250

View Full Product Details